Order Entry
Poland
ContactUsLinkComponent
FEATURES AND BENEFITS INCLUDE: AN EASY-TO-USE BYPASS SWITCH ALLOWS YOU TO TAKE THE ELEGRA OFF-LINE WITHOUT DISCONNECTING ARGON LINES . HIGHLY EFFICIENT MEMBRANE HUMIDIFICATION TECHNOLOGY. IMPROVED SIGNAL STABILITY FOR SAMPLES WITH HIGH TDS. SIMPLE TO 1 * 1 items
Catalog # PERCN8150493
Supplier:  PerkinElmer
FEATURES AND BENEFITS INCLUDE: AN EASY-TO-USE BYPASS SWITCH ALLOWS YOU TO TAKE THE ELEGRA OFF-LINE WITHOUT DISCONNECTING ARGON LINES . HIGHLY EFFICIENT MEMBRANE HUMIDIFICATION TECHNOLOGY. IMPROVED SIGNAL STABILITY FOR SAMPLES WITH HIGH TDS. SIMPLE TO 1 * 1 items
Catalog # PERCN8150493
Supplier:  PerkinElmer
Frequently Bought Together