VWR®, LED-Fluorescence Cubes
Green LED fluorescence cube for TL534B FL, LED emission 523 nm,
Excitation filter 510-550 nm,
Dichroic mirror cut-off 570 nm,
Emission filter 575 LP nm
Excitation filter 510-550 nm,
Dichroic mirror cut-off 570 nm,
Emission filter 575 LP nm
numer katalogowy 630-3246
Dostawca: VWR Collection
Przejdź do:
Product Details & Documents
For use with microscope series TL534B FL.
Documents
VWR® for Microscopy
Product Literature
Dane techniczne
- Description:Green LED fluorescence cube for TL534B FL
- Optical features:LED emission 523 nm,
Excitation filter 510-550 nm,
Dichroic mirror cut-off 570 nm,
Emission filter 575 LP nm



