Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0 zł
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Poland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Hydrochloric Acid 0.02M (0.02N). Duree de conservation : 12 mois. Store under normal laboratory conditions, at temperatures between 15 to 25C 1 * 1 L
Catalog # ACSDZ13414031.1L
Supplier:
CPACHEM
Certificates
Expand image
Hydrochloric Acid 0.02M (0.02N). Duree de conservation : 12 mois. Store under normal laboratory conditions, at temperatures between 15 to 25C 1 * 1 L
Catalog # ACSDZ13414031.1L
Supplier:
CPACHEM
Certificates
Related Information
Documentation
Certificates
Search by Lot Number
Documentation
Frequently Bought Together
VWR® Dosatest, pH indicator test strips, non bleeding
Supplier: VWR Chemicals
pH indicator strips, MQuant®, Supelco®
Supplier: Merck
Ammonia solution ∼32%, GPR RECTAPUR®
Supplier: VWR Chemicals
Test strips, nitrate/nitrite, Dosatest
Supplier: VWR Chemicals
pH indicator papers, MQuant®, Supelco®
Supplier: Merck
Sodium hydroxide 98.5-100.5%, pellets, AnalaR NORMAPUR® Reag. Ph. Eur. analytical reagent
Supplier: VWR Chemicals
Potassium hydroxide 85.0-100.5%, pellets, AnalaR NORMAPUR® Reag. Ph. Eur. analytical reagent
Supplier: VWR Chemicals
UltraClear® for histology, J.T.Baker®
Supplier: Avantor
Test kits, photometric, iron, LCK cuvette tests
Supplier: Hach
Sodium hypochlorite 6…14% (active chlorine) in aqueous solution, GPR RECTAPUR®
Supplier: VWR Chemicals
Sodium hydroxide ≥97.0%, shot, TECHNICAL
Supplier: VWR Chemicals
Test strips for peroxide, MQuant®, Supelco®
Supplier: Merck
Ammonia solution 25%, AnalaR NORMAPUR® analytical reagent
Supplier: VWR Chemicals
Sodium hydroxide, pellets, EMSURE® for analysis, Supelco®
Supplier: Merck
Test Strips, nitrate, MQuant®, Supelco®
Supplier: Merck
Test strips for free chlorine, MQuant®, Supelco®
Supplier: Merck
HPLC columns, NUCLEOSIL® SiOH
Supplier: MACHEREY-NAGEL