Oxalic acid dihydrate AnalaR NORMAPUR analytical reagent R.PE/ACS 1 * 10 kg
Catalog # 20562.410
Supplier: VWR Chemicals
Oxalic acid dihydrate AnalaR NORMAPUR analytical reagent R.PE/ACS 1 * 10 kg
Catalog # 20562.410
Supplier: VWR Chemicals
Related Information
Frequently Bought Together
Sodium chloride ≥99%, GPR RECTAPUR®
Supplier: VWR Chemicals
Sodium dihydrogen phosphate dihydrate ≥99.0%, AnalaR NORMAPUR® analytical reagent
Supplier: VWR Chemicals
(±)-α-Methoxyphenylacetic acid Reag. Ph. Eur., Supelco®
Supplier: Merck
Potassium chloride 99.5-101.0%, AnalaR NORMAPUR® Reag. Ph. Eur. analytical reagent
Supplier: VWR Chemicals
Potassium chloride, EMSURE® for analysis, Supelco®
Supplier: Merck
Ammonium acetate ≥98.0%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Sodium salicylate, EMSURE® for analysis, Supelco®
Supplier: Merck
Potassium hydrogen phthalate ≥99.5%, AnalaR NORMAPUR® analytical reagent
Supplier: VWR Chemicals
Sodium chloride 99.5-100.5%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Sodium chloride regeneration salt, TECHNICAL for water softening
Supplier: VWR Chemicals