Potassium Pyroantimonate TS, Solution 1 * 500 mL
Catalog # SPCMP-198-500ML
Supplier: Spectrum Chemical
Potassium Pyroantimonate TS, Solution 1 * 500 mL
Catalog # SPCMP-198-500ML
Supplier: Spectrum Chemical
Frequently Bought Together
VWR® AVS TITRINORM Buffer Solutions, 20 °C
Supplier: VWR Chemicals
Buffer solutions in sachets, Certipur®, Supelco®
Supplier: Merck
pH buffers, ready to use
Supplier: WTW
VWR®, Bulb Pipettes, One Mark, Class AS
Supplier: VWR Collection
VWR®, Nitrile Gloves
Supplier: VWR Collection
Buffer solutions, Certipur®, colour coded, Supelco®
Supplier: Merck
Acetone ≥99%, TECHNICAL
Supplier: VWR Chemicals
Detergents and cleaning agents, LABWASH® Premium
Supplier: VWR Chemicals
Ammonia solution 28%, AnalaR NORMAPUR® analytical reagent
Supplier: VWR Chemicals
pH buffers, DuraCal
Supplier: HAMILTON BONADUZ
VWR®, Conductivity standard solutions
Supplier: VWR Chemicals