Order Entry
Poland
Orders LinkContactUsLinkComponent
Standard storage cabinets, Protector®

Standard storage cabinets, Protector®

Protector standard storage cabinets support Labconco fume hoods Protector®, with filter panel, Single, left hinge, 610×559×900 mm, 59 kg
Catalog # LBCP9900600
Supplier:   LABCONCO
Standard storage cabinets, Protector®
Catalog # LBCP9900600
Supplier:  LABCONCO
Standard storage cabinets, Protector®
Standard storage cabinets, Protector®

Protector standard storage cabinets support Labconco fume hoods Protector®, with filter panel, Single, left hinge, 610×559×900 mm, 59 kg

Price
Inquire For Price
Log in for availability
1 * 1 SZT
Product Details & Documents

Storage cabinets support fume hoods and used for storage and venting of small laboratory instruments, mild acids and other reagents.

  • Manual-closing, non-locking doors
  • Durable epoxy coated steel construction

 : Includes four leveling feet and one filter panel (20 cm) to increase cabinet depth from 55 to 76,2 cm. Spillstopper™ work surfaces, flat work surfaces, shelf kits and filter panels can be ordered separately.

Specifications
  • Description:
    Protector standard storage cabinets support Labconco fume hoods Protector®, with filter panel
  • Seq:
    06
  • W×D×H:
    610×559×900 mm
  • Doors:
    Single, left hinge
  • Weight:
    59 kg
Accessories

Product Information

  • Catalog #
    Availability
    Price
  • Interior epoxy coated shelf and mounting hardware, 762 mm width, 4,5 kg
    Interior epoxy coated shelf and mounting hardware, 762 mm width, 4,5 kg
    LBCP9919102
    Log in for availability
    1 * 1 SZT