Order Entry
Poland
ContactUsLinkComponent
Human ACE2 / ACEH Protein, His Tag Human ACE2, His Tag (AC2-H52H8) is expressed from human 293 cells (HEK293). It contains AA Gln 18 - Ser 740 (Accession Q9BYF1-1). This protein carries a polyhistidine tag at the C-terminus. The protein has a calc .5 1 * 50 µG
Catalog # ACRBAC2H52H850UG
Supplier:  ACRO Biosystems
Human ACE2 / ACEH Protein, His Tag Human ACE2, His Tag (AC2-H52H8) is expressed from human 293 cells (HEK293). It contains AA Gln 18 - Ser 740 (Accession Q9BYF1-1). This protein carries a polyhistidine tag at the C-terminus. The protein has a calc .5 1 * 50 µG
Catalog # ACRBAC2H52H850UG
Supplier:  ACRO Biosystems
Frequently Bought Together