Order Entry
Poland
ContactUsLinkComponent
Cynomolgus ACE2 / ACEH Protein, His Tag Cynomolgus ACE2, His Tag (AC2-C52H7) is expressed from human 293 cells (HEK293). It contains AA Gln 18 - Thr 741 (Accession A0A2K5X283-1). This protein carries a polyhistidine tag at the C-terminus. The prot la 1 * 1 mg
Catalog # ACRBAC2C52H71MG
Supplier:  ACRO Biosystems
Cynomolgus ACE2 / ACEH Protein, His Tag Cynomolgus ACE2, His Tag (AC2-C52H7) is expressed from human 293 cells (HEK293). It contains AA Gln 18 - Thr 741 (Accession A0A2K5X283-1). This protein carries a polyhistidine tag at the C-terminus. The prot la 1 * 1 mg
Catalog # ACRBAC2C52H71MG
Supplier:  ACRO Biosystems
Frequently Bought Together