Order Entry
Poland
ContactUsLinkComponent
Rat ACE2 / ACEH Protein, His Tag Rat ACE2, His Tag (AC2-R5246) is expressed from human 293 cells (HEK293). It contains AA Gln 18 - T hr 740 (Accession Q5EGZ1-1). This protein carries a polyhistidine tag at the N-terminus. The protein has a calculat D 1 * 50 µG
Catalog # ACRBAC2R524650UG
Dostawca:  ACRO Biosystems
Rat ACE2 / ACEH Protein, His Tag Rat ACE2, His Tag (AC2-R5246) is expressed from human 293 cells (HEK293). It contains AA Gln 18 - T hr 740 (Accession Q5EGZ1-1). This protein carries a polyhistidine tag at the N-terminus. The protein has a calculat D 1 * 50 µG
Catalog # ACRBAC2R524650UG
Dostawca:  ACRO Biosystems
Frequently Bought Together