Order Entry
Poland
ContactUsLinkComponent
Mouse ACE2 / ACEH Protein, His Tag Mouse ACE2, His Tag (AC2-M5248) is expressed from human 293 cells (HEK293). It contains AA Gln 18 - Thr 740 (Accession Q8R0I0-1). This protein carries a polyhistidine tag at the N-terminus. The protein has a calc .5 1 * 50 µG
  ACRBAC2M524850UG
Mouse ACE2 / ACEH Protein, His Tag Mouse ACE2, His Tag (AC2-M5248) is expressed from human 293 cells (HEK293). It contains AA Gln 18 - Thr 740 (Accession Q8R0I0-1). This protein carries a polyhistidine tag at the N-terminus. The protein has a calc .5 1 * 50 µG
  ACRBAC2M524850UG
Frequently Bought Together