Order Entry
Poland
ContactUsLinkComponent
2019-nCoV (COVID-19) S protein RBD, Mouse IgG2a Fc Tag 2019-nCoV (COVID-19) S protein RBD, Mouse IgG2a Fc Tag (SPD-C5259) is express ed from human 293 cells (HEK293). It contains AA Arg 319 - Phe 541 (Accession QHD43416.1). This protein carries a m a 1 * 100 µG
Catalog # ACRBSPDC5259100UG
Supplier:  ACRO Biosystems
2019-nCoV (COVID-19) S protein RBD, Mouse IgG2a Fc Tag 2019-nCoV (COVID-19) S protein RBD, Mouse IgG2a Fc Tag (SPD-C5259) is express ed from human 293 cells (HEK293). It contains AA Arg 319 - Phe 541 (Accession QHD43416.1). This protein carries a m a 1 * 100 µG
Catalog # ACRBSPDC5259100UG
Supplier:  ACRO Biosystems
Frequently Bought Together