Order Entry
Poland
ContactUsLinkComponent
2019-nCoV (COVID-19) S1 protein, Mouse IgG2a Fc Tag 2019-nCoV (COVID-19) S1 protein, Mouse IgG2a Fc Tag (S1N-C5257) is expressed fro m human 293 cells (HEK293). It contains AA Val 16 - Arg 685 (Accession QHD43416.1). This protein carries a mouse Ig h 1 * 100 µG
Catalog # ACRBS1NC5257100UG
Supplier:  ACROBIOSYSTEMS
2019-nCoV (COVID-19) S1 protein, Mouse IgG2a Fc Tag 2019-nCoV (COVID-19) S1 protein, Mouse IgG2a Fc Tag (S1N-C5257) is expressed fro m human 293 cells (HEK293). It contains AA Val 16 - Arg 685 (Accession QHD43416.1). This protein carries a mouse Ig h 1 * 100 µG
Catalog # ACRBS1NC5257100UG
Supplier:  ACROBIOSYSTEMS
Frequently Bought Together