[EN]1-[3-(DIMETHYLAMINO)PROPYL]-3-ETHYLU 1 * 5 g
Catalog # CAROFE30152-5G
Supplier: CARBOSYNTH
Frequently Bought Together
Methanol, Prepsolv® for preparative chromatography, Supelco®
Supplier: Merck
n-Hexane, LiChrosolv® for liquid chromatography, Supelco®
Supplier: Merck
Diethyl ether, SupraSolv® for gas chromatography, Supelco®
Supplier: Merck
2-Propanol ≥98%, TECHNICAL
Supplier: VWR Chemicals
Chloroform, LiChrosolv® for HPLC
Supplier: Merck
Methanol ≥99.5% NF, Ph. Eur., BP
Supplier: VWR Chemicals
Tetrachloroethylene 99%, extra pure
Supplier: Thermo Fisher Scientific
Transport trolleys, BC series
Supplier: Metro International
Ethylene glycol ≥99%
Supplier: Thermo Fisher Scientific
(±)-2-Methyltetrahydrofuran, anhydrous stabilised for grignard-reaction
Supplier: Thermo Fisher Scientific
Ethanol absolute ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Acetone ≥99.5% (by GC), Puriss. p.a. ACS, ISO, Reag. Ph. Eur., Riedel-de Haën™
Supplier: Honeywell Chemicals