Order Entry
Poland
Orders LinkContactUsLinkComponent
Polyoxylglycerides, Reference Standards
Polyoxylglycerides, Reference Standards
Lauroyl Polyoxylglycerides 300, USP reference standard, Ampule, 500 mg
Catalog # 1356993.
Supplier:   USP
Polyoxylglycerides, Reference Standards
Catalog # 1356993.
Supplier:  USP
CAS Number:  
Polyoxylglycerides, Reference Standards
Lauroyl Polyoxylglycerides 300, USP reference standard, Ampule, 500 mg
CAS Number:  
Price
2 130 zł
Log in for availability
1 * 500 mg
Product Details & Documents
USP Reference Standards are specified for use in conducting official USP–NF tests and assays. USP also provides Reference Standards specified in the Food Chemicals Codex as well as authentic substances—high-quality chemical samples—as a service to analytical, clinical, pharmaceutical and research laboratories. To confirm accuracy and reproducibility, USP Reference Standards are rigorously tested and evaluated by multiple independent laboratories including USP, commercial, regulatory, and academic labs. USP also provide publicly available, official documentary standards for pharmaceutical ingredients in the USP–NF that link directly with our primary reference standards.
Certifications: US Pharmacopea Reference Standard
Specifications
Specifications
  • Pack type:
    Ampule
  • Pk:
    500 mg
  • Standards:
    USP reference standard
  • Description:
    Lauroyl Polyoxylglycerides 300
  • Cat. No.:
    1356993.
  • Description:
    Lauroyl Polyoxylglycerides 300
  • CAS No.:
    12737-91-6