Iodomethane, 99%, stab. with copper 1 * 250 g
Catalog # ALFAA10559.30
Supplier: Thermo Fisher Scientific
Iodomethane, 99%, stab. with copper 1 * 250 g
Catalog # ALFAA10559.30
Supplier: Thermo Fisher Scientific
Frequently Bought Together
Tetraheptylammonium bromide, Supelco®
Supplier: Merck
(±)-2-Methyltetrahydrofuran 99+% stabilised, pure
Supplier: Thermo Fisher Scientific
Toluene, pure analytical reagent
Supplier: CHEMPUR
Ethyl acetate ≥99.5%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
N,N-Dimethylformamide, pure analytical reagent
Supplier: CHEMPUR
2-Propanol, extra dry 99.8%, AcroSeal®
Supplier: Thermo Fisher Scientific
Hydrochloric acid ≥36.5% ≥99.999% (metals basis)
Supplier: Thermo Fisher Scientific
Pentane reagent grade, 98% 1 * 1 L
Supplier: Merck
1-Butanol ≥99.7%, CHROMASOLV™ Plus for HPLC, Riedel-de Haën™
Supplier: Honeywell Chemicals
(±)-1,2-Propanediol, pure
Supplier: CHEMPUR
Isoamyl Alcohol according to Gerber for analysis 1 * 1.000 mL
Supplier: PanReac AppliChem
Ethanolamine, Sigma-Aldrich®
Supplier: Merck
Tetrabutylammonium hydroxide 0.4 M in water for HPLC
Supplier: Thermo Fisher Scientific
Methanol ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
1,4-Dioxane, extra dry over Molecular Sieve 99,5% stabilised, AcroSeal®
Supplier: Thermo Fisher Scientific
Chloroform ≥99.5% stabilised
Supplier: Honeywell Chemicals