Cynomolgus B7-H3, His Tag (B73-C52Ha) is expressed from human 293 cells (HEK293). It contains AA Leu 29 - Glu 465 (Accession XP01530 8534.1). 1 * 100 µG
Cynomolgus B7-H3, His Tag (B73-C52Ha) is expressed from human 293 cells (HEK293). It contains AA Leu 29 - Glu 465 (Accession XP01530 8534.1). 1 * 100 µG