Order Entry
Poland
ContactUsLinkComponent
Recombinat Protein,Mouse CD24 Protein, His Tag,Mouse CD24, His Tag (CD4-M52H7) is expressed from human 293 cells (HEK293). It contai ns AA Asn 27 - Gly 53 (Accession P24807-1).This protein carries a polyhistidine tag at the C-terminus. The protein ha 1 * 1 mg
Catalog # ACRBCD4-M52H7-1MG
Supplier:  ACRO Biosystems
Recombinat Protein,Mouse CD24 Protein, His Tag,Mouse CD24, His Tag (CD4-M52H7) is expressed from human 293 cells (HEK293). It contai ns AA Asn 27 - Gly 53 (Accession P24807-1).This protein carries a polyhistidine tag at the C-terminus. The protein ha 1 * 1 mg
Catalog # ACRBCD4-M52H7-1MG
Supplier:  ACRO Biosystems
Frequently Bought Together