Order Entry
Poland
ContactUsLinkComponent
Recombinat Protein,Cynomolgus Siglec-9 Protein, His Tag,Cynomolgus Siglec-9, His Tag (SI9-C52H9) is expressed from human 293 cells ( HEK293). It contains AA Gln 20 - Gly 350 (Accession XP005590140.1).This protein carries a polyhistidine tag at the C- 1 * 100 µG
Catalog # ACRBSI9-C52H9100UG
Supplier:  ACRO Biosystems
Recombinat Protein,Cynomolgus Siglec-9 Protein, His Tag,Cynomolgus Siglec-9, His Tag (SI9-C52H9) is expressed from human 293 cells ( HEK293). It contains AA Gln 20 - Gly 350 (Accession XP005590140.1).This protein carries a polyhistidine tag at the C- 1 * 100 µG
Catalog # ACRBSI9-C52H9100UG
Supplier:  ACRO Biosystems
Frequently Bought Together