Order Entry
Poland
ContactUsLinkComponent
Recombinat Protein,Human CD300LG Nepmucin Protein, His Tag,Human CD300LG, His Tag (CDG-H5223) is expressed from human 293 cells (HEK 293). It contains AA Leu 19 - Arg 247 (Accession AAH25395).This protein carries a polyhistidine tag at the C-terminus 1 * 1 mg
  ACRBCDG-H5223-1MG
Recombinat Protein,Human CD300LG Nepmucin Protein, His Tag,Human CD300LG, His Tag (CDG-H5223) is expressed from human 293 cells (HEK 293). It contains AA Leu 19 - Arg 247 (Accession AAH25395).This protein carries a polyhistidine tag at the C-terminus 1 * 1 mg
  ACRBCDG-H5223-1MG
Frequently Bought Together