Recombinat Protein,Human CD5 Protein, His Tag,Human CD5, His Tag (CD5-H52H5) is expressed from human 293 cells (HEK293). It contains AA Arg 25 - Pro 372 (Accession NP055022).This protein carries a polyhistidine tag at the C-terminus. The protein has 1 * 100 µG
Catalog # ACRBCD5-H52H5100UG
Supplier: ACRO Biosystems
Frequently Bought Together