Order Entry
Poland
ContactUsLinkComponent
Recombinat Protein,Human CG alpha CGA Protein, His Tag,Human CG alpha, His Tag (CGA-H52H2) is expressed from human 293 cells (HEK293 ). It contains AA Ala 25 - Ser 116 (Accession AAH20782).This protein carries a polyhistidine tag at the C-terminus. T 1 * 1 mg
Catalog # ACRBCGA-H52H2-1MG
Supplier:  ACRO Biosystems
Recombinat Protein,Human CG alpha CGA Protein, His Tag,Human CG alpha, His Tag (CGA-H52H2) is expressed from human 293 cells (HEK293 ). It contains AA Ala 25 - Ser 116 (Accession AAH20782).This protein carries a polyhistidine tag at the C-terminus. T 1 * 1 mg
Catalog # ACRBCGA-H52H2-1MG
Supplier:  ACRO Biosystems
Frequently Bought Together