Order Entry
Poland
ContactUsLinkComponent
Recombinat Protein,Human Cystatin S CST4 Protein, His Tag,Human Cystatin S, His Tag (CS4-H5223) is expressed from human 293 cells (H EK293). It contains AA Ser 21 - Ala 141 (Accession AAH65714).This protein carries a polyhistidine tag at the C-termin 1 * 100 µG
Catalog # ACRBCS4-H5223100UG
Supplier:  ACRO Biosystems
Recombinat Protein,Human Cystatin S CST4 Protein, His Tag,Human Cystatin S, His Tag (CS4-H5223) is expressed from human 293 cells (H EK293). It contains AA Ser 21 - Ala 141 (Accession AAH65714).This protein carries a polyhistidine tag at the C-termin 1 * 100 µG
Catalog # ACRBCS4-H5223100UG
Supplier:  ACRO Biosystems
Frequently Bought Together