Order Entry
Poland
ContactUsLinkComponent
ActiveMax® Human Activin A, Tag Free, low endotoxin (ACA-H421b) is expressed from human 293 cells (HEK293). It contains AA Gly 311 - Ser 426 (Accession AAH07858.1). 1 * 1 mg
Catalog # ACRBACA-H421B-1MG
Supplier:  ACRO Biosystems
ActiveMax® Human Activin A, Tag Free, low endotoxin (ACA-H421b) is expressed from human 293 cells (HEK293). It contains AA Gly 311 - Ser 426 (Accession AAH07858.1). 1 * 1 mg
Catalog # ACRBACA-H421B-1MG
Supplier:  ACRO Biosystems
Frequently Bought Together