Recombinat Protein,Human EMMPRIN CD147 Protein, His Tag,Human EMMPRIN, His Tag (CD7-H5222) is expressed from human 293 cells (HEK293 ). It contains AA Ala 22 - His 205 (Accession NP940991.1).This protein carries a polyhistidine tag at the C-terminus. 1 * 100 µG
Catalog # ACRBCD7-H5222100UG
Supplier: ACRO Biosystems
Frequently Bought Together