Recombinat Protein,Human Ephrin-A3 EFNA3 Protein, Fc Tag,Human Ephrin-A3, Fc Tag (EA3-H5258) is expressed from human 293 cells (HEK2 93). It contains AA Gln 23 - Ser 213 (Accession AAI10407).This protein carries a human IgG1 Fc tag at the C-terminus. 1 * 1 mg
Catalog # ACRBEA3-H5258-1MG
Supplier: ACROBIOSYSTEMS
Frequently Bought Together