Order Entry
Poland
ContactUsLinkComponent
ActiveMax® Human IFN-gamma, Tag Free (IFG-H4211) is expressed from human 293 cells (HEK293). It contains AA Gln 24 - Gln 166 (Access ion AAH70256). 1 * 1 mg
Catalog # ACRBIFG-H4211-1MG
Dostawca:  ACRO Biosystems
ActiveMax® Human IFN-gamma, Tag Free (IFG-H4211) is expressed from human 293 cells (HEK293). It contains AA Gln 24 - Gln 166 (Access ion AAH70256). 1 * 1 mg
Catalog # ACRBIFG-H4211-1MG
Dostawca:  ACRO Biosystems
Frequently Bought Together