Recombinat Protein,Human E-Selectin CD62E Protein, His Tag,Human E-Selectin, His Tag (CDE-H5222) is expressed from human 293 cells ( HEK293). It contains AA Trp 22 - Pro 556 (Accession NP000441.2).This protein carries a polyhistidine tag at the C-ter 1 * 200 µG
Catalog # ACRBCDE-H5222200UG
Supplier: ACRO Biosystems
Frequently Bought Together