Order Entry
Poland
ContactUsLinkComponent
MABSol® Biotinylated Human GHR, His Tag, ultra sensitivity (primary amine labeling) (GHR-H8222) is expressed from human HEK293 cells . It contains AA Ala 27 - Tyr 264 (Accession P10912-1). It is the biotinylated form of Human Growth Hormone R (GHR) P 1 * 50 µG
Catalog # ACRBGHR-H8222-50UG
Supplier:  ACROBIOSYSTEMS
MABSol® Biotinylated Human GHR, His Tag, ultra sensitivity (primary amine labeling) (GHR-H8222) is expressed from human HEK293 cells . It contains AA Ala 27 - Tyr 264 (Accession P10912-1). It is the biotinylated form of Human Growth Hormone R (GHR) P 1 * 50 µG
Catalog # ACRBGHR-H8222-50UG
Supplier:  ACROBIOSYSTEMS
Frequently Bought Together