Order Entry
Poland
ContactUsLinkComponent
MABSol® Biotinylated Human PCSK9, Avitag,His Tag (PC9-H82E7) is expressed from human HEK293 cells. It contains AA Gln 31 - Gln 692 (Accession Q8NBP7-1). null
Catalog # ACRBPC9-H82E7200UG
Supplier:  ACRO Biosystems
MABSol® Biotinylated Human PCSK9, Avitag,His Tag (PC9-H82E7) is expressed from human HEK293 cells. It contains AA Gln 31 - Gln 692 (Accession Q8NBP7-1). null
Catalog # ACRBPC9-H82E7200UG
Supplier:  ACRO Biosystems
Frequently Bought Together