Order Entry
Poland
ContactUsLinkComponent
Biotinylated Human EGF R, His,Avitag (EGR-H82E3) is expressed from human 293 cells (HEK293). It contains AA Leu 25 - Ser 645 (Access ion P00533-1). 1 * 200 µG
Catalog # ACRBEGR-H82E3200UG
Supplier:  ACROBIOSYSTEMS
Biotinylated Human EGF R, His,Avitag (EGR-H82E3) is expressed from human 293 cells (HEK293). It contains AA Leu 25 - Ser 645 (Access ion P00533-1). 1 * 200 µG
Catalog # ACRBEGR-H82E3200UG
Supplier:  ACROBIOSYSTEMS
Frequently Bought Together