Order Entry
Poland
ContactUsLinkComponent
MABSol® Biotinylated Human TFPI, His Tag, primary amine labeling (TFI-H8226) is expressed from human HEK293 cells. It contains AA A sp 29 - Lys 282 (Accession NP006278.1). It is the biotinylated form of Human TFPI, His Tag (TFI-H5226). null
Catalog # ACRBTFI-H8226200UG
Supplier:  ACRO Biosystems
MABSol® Biotinylated Human TFPI, His Tag, primary amine labeling (TFI-H8226) is expressed from human HEK293 cells. It contains AA A sp 29 - Lys 282 (Accession NP006278.1). It is the biotinylated form of Human TFPI, His Tag (TFI-H5226). null
Catalog # ACRBTFI-H8226200UG
Supplier:  ACRO Biosystems
Frequently Bought Together