Biotinylated Human Mesothelin (296-580), His,Avitag (MSN-H82E9) is expressed from human 293 cells (HEK293). It contains AA Glu 296 - Gly 580 (Accession AAH09272.1). 1 * 200 µG
Catalog # ACRBMSN-H82E9200UG
Supplier: ACRO Biosystems
Frequently Bought Together