Order Entry
Poland
ContactUsLinkComponent
Cynomolgus CD40, His Tag (CD0-C52H6) is expressed from human 293 cells (HEK293). It contains AA Glu 21 - Arg 193 (Accession G7PG38). 1 * 100 µG
Catalog # ACRBCD0-C52H6100UG
Supplier:  ACRO Biosystems
Cynomolgus CD40, His Tag (CD0-C52H6) is expressed from human 293 cells (HEK293). It contains AA Glu 21 - Arg 193 (Accession G7PG38). 1 * 100 µG
Catalog # ACRBCD0-C52H6100UG
Supplier:  ACRO Biosystems
Frequently Bought Together