Cynomolgus Rhesus macaque CTLA-4, His Tag (CT4-C5227) is expressed from human 293 cells (HEK293). It contains AA Ala 37 - Ser 160 (A ccession G7PL88-1). In the region Ala 37 - Ser 160, the AA sequence of Cynomolgus and Rhesus macaque CTLA-4 are homol 1 * 200 µG
Catalog # ACRBCT4-C5227200UG
Supplier: ACRO Biosystems
Frequently Bought Together