Order Entry
Poland
ContactUsLinkComponent
Recombinat Protein,Cynomolgus DR3 TNFRSF25 Protein, Fc Tag,Cynomolgus DR3, Fc Tag (TN5-C5257) is expressed from human 293 cells (HEK 293). It contains AA Gln 20 - Gly 203 (Accession G7NTB5).This protein carries a human IgG1 Fc tag at the C-terminus. 1 * 100 µG
Catalog # ACRBTN5-C5257100UG
Supplier:  ACROBIOSYSTEMS
Recombinat Protein,Cynomolgus DR3 TNFRSF25 Protein, Fc Tag,Cynomolgus DR3, Fc Tag (TN5-C5257) is expressed from human 293 cells (HEK 293). It contains AA Gln 20 - Gly 203 (Accession G7NTB5).This protein carries a human IgG1 Fc tag at the C-terminus. 1 * 100 µG
Catalog # ACRBTN5-C5257100UG
Supplier:  ACROBIOSYSTEMS
Frequently Bought Together