Order Entry
Poland
ContactUsLinkComponent
Human TNFSF11, His Tag (active trimer) (RAL-H5240) is expressed from human 293 cells (HEK293). It contains AA Gly 64 - Asp 245 (Acce ssion AAC51762.1). 1 * 50 µG
Catalog # ACRBRAL-H5240-50UG
Supplier:  ACRO Biosystems
Human TNFSF11, His Tag (active trimer) (RAL-H5240) is expressed from human 293 cells (HEK293). It contains AA Gly 64 - Asp 245 (Acce ssion AAC51762.1). 1 * 50 µG
Catalog # ACRBRAL-H5240-50UG
Supplier:  ACRO Biosystems
Frequently Bought Together