Order Entry
Poland
ContactUsLinkComponent
Cynomolgus Rhesus macaque TIGIT, Fc Tag (TIT-C5254) is expressed from human 293 cells (HEK293). It contains AA Met 89 - Ile 208 (Acc ession G7NXM4-1). In the region Met 89 - Ile 208, the AA sequence of Cynomolgus and Rhesus macaque TIGIT are homologu 1 * 1 mg
Catalog # ACRBTIT-C5254-1MG
Supplier:  ACRO Biosystems
Cynomolgus Rhesus macaque TIGIT, Fc Tag (TIT-C5254) is expressed from human 293 cells (HEK293). It contains AA Met 89 - Ile 208 (Acc ession G7NXM4-1). In the region Met 89 - Ile 208, the AA sequence of Cynomolgus and Rhesus macaque TIGIT are homologu 1 * 1 mg
Catalog # ACRBTIT-C5254-1MG
Supplier:  ACRO Biosystems
Frequently Bought Together