Recombinat Protein,Mouse LILRB4 CD85k ILT3 Protein, Fc Tag,Mouse LILRB4, Fc Tag (CDK-M5250) is expressed from human 293 cells (HEK29 3). It contains AA Gly 24 - Lys 238 (Accession Q64281-1).This protein carries a human IgG1 Fc tag at the C-terminus. 1 * 1 mg
Catalog # ACRBCDK-M5250-1MG
Supplier: ACRO Biosystems
Frequently Bought Together