Order Entry
Poland
ContactUsLinkComponent
Biotinylated Mouse FOLR1, His,Avitag (FO1-M82E9) is expressed from human 293 cells (HEK293). It contains AA Thr 25 - Ser 232 (Access ion P35846-1). 1 * 25 µG
Catalog # ACRBFO1-M82E9-25UG
Supplier:  ACRO Biosystems
Biotinylated Mouse FOLR1, His,Avitag (FO1-M82E9) is expressed from human 293 cells (HEK293). It contains AA Thr 25 - Ser 232 (Access ion P35846-1). 1 * 25 µG
Catalog # ACRBFO1-M82E9-25UG
Supplier:  ACRO Biosystems
Frequently Bought Together