Order Entry
Poland
ContactUsLinkComponent
Human B7-H2, His Tag (B72-H5221) is expressed from human 293 cells (HEK293). It contains AA Asp 19 - Ser 258 (Accession NP056074). 1 * 200 µG
Catalog # ACRBB72-H5221200UG
Supplier:  ACRO Biosystems
Human B7-H2, His Tag (B72-H5221) is expressed from human 293 cells (HEK293). It contains AA Asp 19 - Ser 258 (Accession NP056074). 1 * 200 µG
Catalog # ACRBB72-H5221200UG
Supplier:  ACRO Biosystems
Frequently Bought Together