Order Entry
Poland
ContactUsLinkComponent
Human B7-H4, His Tag (B74-H5222) is expressed from human 293 cells (HEK293). It contains AA Phe 29 - Ala 258 (Accession NP078902). 1 * 100 µG
Catalog # ACRBB74-H5222100UG
Supplier:  ACRO Biosystems
Human B7-H4, His Tag (B74-H5222) is expressed from human 293 cells (HEK293). It contains AA Phe 29 - Ala 258 (Accession NP078902). 1 * 100 µG
Catalog # ACRBB74-H5222100UG
Supplier:  ACRO Biosystems
Frequently Bought Together