Order Entry
Poland
ContactUsLinkComponent
Recombinat Protein,Human Betacellulin Protein, Fc Tag,Human Betacellulin, Fc Tag (BEN-H525x) is expressed from human 293 cells (HEK2 93). It contains AA Asp 32 - Tyr 111 (Accession AAH11618).This protein carries a human IgG1 Fc tag at the C-terminus. 1 * 50 µG
Catalog # ACRBBEN-H525X-50UG
Supplier:  ACRO Biosystems
Recombinat Protein,Human Betacellulin Protein, Fc Tag,Human Betacellulin, Fc Tag (BEN-H525x) is expressed from human 293 cells (HEK2 93). It contains AA Asp 32 - Tyr 111 (Accession AAH11618).This protein carries a human IgG1 Fc tag at the C-terminus. 1 * 50 µG
Catalog # ACRBBEN-H525X-50UG
Supplier:  ACRO Biosystems
Frequently Bought Together