Order Entry
Poland
ContactUsLinkComponent
Human BMP-2, Tag Free (BM2-H4117) is expressed from E.coli cells. It contains AA Ala 284 - Arg 396 (Accession NP001191). 1 * 200 µG
Catalog # ACRBBM2-H4117200UG
Supplier:  ACRO Biosystems
Human BMP-2, Tag Free (BM2-H4117) is expressed from E.coli cells. It contains AA Ala 284 - Arg 396 (Accession NP001191). 1 * 200 µG
Catalog # ACRBBM2-H4117200UG
Supplier:  ACRO Biosystems
Frequently Bought Together