Order Entry
Poland
ContactUsLinkComponent
Recombinat Protein,Human BMPR-IA ALK-3 Protein, Fc Tag,Human BMPR-IA, Fc Tag (AL3-H5259) is expressed from human 293 cells (HEK293). It contains AA Gln 24 - Arg 152 (Accession CAA80257).This protein carries a human IgG1 Fc tag at the C-terminus. The 1 * 200 µG
Catalog # ACRBAL3-H5259200UG
Supplier:  ACROBIOSYSTEMS
Recombinat Protein,Human BMPR-IA ALK-3 Protein, Fc Tag,Human BMPR-IA, Fc Tag (AL3-H5259) is expressed from human 293 cells (HEK293). It contains AA Gln 24 - Arg 152 (Accession CAA80257).This protein carries a human IgG1 Fc tag at the C-terminus. The 1 * 200 µG
Catalog # ACRBAL3-H5259200UG
Supplier:  ACROBIOSYSTEMS
Frequently Bought Together