Order Entry
Poland
ContactUsLinkComponent
Cynomolgus LDL R, His Tag (LDR-C5222) is expressed from human 293 cells (HEK293). It contains AA Ala 22 - Gly 788 (Accession XP00558 8053.1). 1 * 100 µG
Catalog # ACRBLDR-C5222100UG
Dostawca:  ACRO Biosystems
Cynomolgus LDL R, His Tag (LDR-C5222) is expressed from human 293 cells (HEK293). It contains AA Ala 22 - Gly 788 (Accession XP00558 8053.1). 1 * 100 µG
Catalog # ACRBLDR-C5222100UG
Dostawca:  ACRO Biosystems
Frequently Bought Together