Recombinat Protein,Human Carbonic Anhydrase IX CA9 (138-414) Protein, His Tag,Human Carbonic Anhydrase IX (138-414), His Tag (CA9-H5 220) is expressed from human 293 cells (HEK293). It contains AA Gln 138 - Asp 414 (Accession NP001207.2).This protein 1 * 50 µG
Catalog # ACRBCA9-H5220-50UG
Supplier: ACRO Biosystems
Frequently Bought Together