Order Entry
Poland
ContactUsLinkComponent
Biotinylated Mouse CD155, Fc,Avitag (CD5-M82F7) is expressed from human 293 cells (HEK293). It contains AA Asp 29 - Leu 348 (Accessi on Q8K094-1). 1 * 25 µG
Catalog # ACRBCD5-M82F7-25UG
Supplier:  ACRO Biosystems
Biotinylated Mouse CD155, Fc,Avitag (CD5-M82F7) is expressed from human 293 cells (HEK293). It contains AA Asp 29 - Leu 348 (Accessi on Q8K094-1). 1 * 25 µG
Catalog # ACRBCD5-M82F7-25UG
Supplier:  ACRO Biosystems
Frequently Bought Together