Recombinat Protein,Human HE4 WFDC2 Protein, His Tag,Human HE4, His Tag (HE4-H5223) is expressed from human 293 cells (HEK293). It co ntains AA Glu 31 - Phe 124 (Accession AAH46106).This protein carries a polyhistidine tag at the C-terminus. The prote 1 * 1 mg
Catalog # ACRBHE4-H5223-1MG
Supplier: ACRO Biosystems
Frequently Bought Together