Order Entry
Poland
ContactUsLinkComponent
Human TGFBR2, Fc Tag (TG2-H5252) is expressed from human 293 cells (HEK293). It contains AA Thr 23 - Asp 159 (Accession NP003233). 1 * 100 µG
Catalog # ACRBTG2-H5252100UG
Supplier:  ACRO Biosystems
Human TGFBR2, Fc Tag (TG2-H5252) is expressed from human 293 cells (HEK293). It contains AA Thr 23 - Asp 159 (Accession NP003233). 1 * 100 µG
Catalog # ACRBTG2-H5252100UG
Supplier:  ACRO Biosystems
Frequently Bought Together