Order Entry
Poland
ContactUsLinkComponent
Recombinat Protein,Human Thrombospondin-2 Protein, His Tag,Human Thrombospondin-2, His Tag (TH2-H52H5) is expressed from human 293 c ells (HEK293). It contains AA Gly 19 - Ile 1172 (Accession P35442-1).This protein carries a polyhistidine tag at the 1 * 100 µG
Catalog # ACRBTH2-H52H5100UG
Supplier:  ACRO Biosystems
Recombinat Protein,Human Thrombospondin-2 Protein, His Tag,Human Thrombospondin-2, His Tag (TH2-H52H5) is expressed from human 293 c ells (HEK293). It contains AA Gly 19 - Ile 1172 (Accession P35442-1).This protein carries a polyhistidine tag at the 1 * 100 µG
Catalog # ACRBTH2-H52H5100UG
Supplier:  ACRO Biosystems
Frequently Bought Together